siRNA Details
| Gene Symbol: Sept7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205367.1 | NP_001192296.1 | 2 | 347 | ||
| NM_009859.4 | NP_033989.2 | 3 | 350 | |||
| GenBank mRNA | AK088997.1 | - | 3 | 137 | ||
| AK141572.1 | - | 3 | 142 | |||
| AK144127.1 | - | 3 | 139 | |||
| AK147986.1 | - | 3 | 142 | |||
| AK220447.1 | - | 2 | 120 | |||
| BC058587.1 | - | 2 | 151 | 6529169 | ||

Sept7
Genomic Map