siRNA Details
| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141737.1 | - | 3 | 438 | ||
| AK168287.1 | - | 3 | 433 | |||
| BC004669.1 | - | 2 | 142 | 3498846 | ||
| L16926.1 | - | 2 | 566 | |||
| U15562.1 | - | 3 | 404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc25c| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141737.1 | - | 3 | 438 | ||
| AK168287.1 | - | 3 | 433 | |||
| BC004669.1 | - | 2 | 142 | 3498846 | ||
| L16926.1 | - | 2 | 566 | |||
| U15562.1 | - | 3 | 404 | |||