siRNA Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB018574.1 | - | 3 | 315 | ||
| AB018575.1 | - | 3 | 315 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc7| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB018574.1 | - | 3 | 315 | ||
| AB018575.1 | - | 3 | 315 | |||