siRNA Details
| Gene Symbol: Capzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037761.2 | NP_001032850.1 | 3 | 306 | ||
| NM_001271405.1 | NP_001258334.1 | 3 | 285 | |||
| NM_001271406.1 | NP_001258335.1 | 3 | 271 | |||
| NM_009798.4 | NP_033928.1 | 3 | 306 | |||
| GenBank mRNA | AK153367.1 | - | 3 | 237 | ||
| AK153394.1 | - | 3 | 237 | |||
| AK156778.1 | - | 3 | 275 | |||
| AK159390.1 | - | 3 | 280 | |||
| AK160082.1 | - | 3 | 281 | |||
| AK162757.1 | - | 3 | 273 | |||
| AK168738.1 | - | 3 | 234 | |||
| BC002053.1 | - | 3 | 248 | 3590980 | ||
| BY033332.1 | - | 3 | 306 | |||
| BY124858.1 | - | 3 | 295 | |||
| FJ692320.1 | - | 3 | 271 | |||
| U10406.1 | - | 3 | 226 | |||
| U10407.1 | - | 3 | 197 | |||

Capzb
Genomic Map