siRNA Details
| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141737.1 | - | 4, 5 | 640 | ||
| AK168287.1 | - | 4, 5 | 635 | |||
| AK181318.1 | - | 1, 2 | 34 | |||
| BC004669.1 | - | 3, 4 | 344 | 3498846 | ||
| L16926.1 | - | 3, 4 | 768 | |||
| U15562.1 | - | 4, 5 | 606 | |||

Cdc25c
Genomic Map