siRNA Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009794.3 | NP_033924.2 | 21 | 2542 | ||
| GenBank mRNA | AF015038.1 | - | 21 | 2488 | ||
| AK052141.1 | - | 19 | 2131 | |||
| AK150695.1 | - | 21 | 2516 | |||
| AK152094.1 | - | 21 | 2516 | |||
| AK152790.1 | - | 21 | 2519 | |||
| AK152994.1 | - | 21 | 2519 | |||
| AK154855.1 | - | 21 | 2556 | |||
| AK219579.1 | - | 1 | 269 | |||
| BC054726.1 | - | 21 | 2542 | 5703563 | ||
| D38117.1 | - | 21 | 2583 | |||

Capn2
Genomic Map