siRNA Details
| Gene Symbol: Cd24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009846.2 | NP_033976.1 | 2 | 498 | ||
| GenBank mRNA | AK014279.1 | - | 2 | 498 | ||
| AK133869.1 | - | 2 | 497 | |||
| AK168111.1 | - | 2 | 497 | |||
| AK169613.1 | - | 2 | 495 | |||
| BC023197.1 | - | 1 | 222 | |||
| BC026702.1 | - | 2 | 509 | |||
| BC075622.1 | - | 2 | 478 | 6415159 | ||
| M58661.1 | - | 2 | 485 | |||
| X53825.1 | - | 2 | 436 | |||

Cd24a
Genomic Map