siRNA Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037801.2 | NP_001032890.1 | 8 | 1546 | ||
| NM_009852.3 | NP_033982.3 | 8 | 1546 | |||
| GenBank mRNA | AK030822.1 | - | 8 | 1516 | ||
| AK030823.1 | - | 8 | 1547 | |||
| AK088050.1 | - | 8 | 1524 | |||
| AK141213.1 | - | 4 | 693 | |||
| BC014294.1 | - | 8 | 1552 | 4187689 | ||
| U12434.1 | - | 8 | 1344 | |||
| U35370.1 | - | 8 | 1462 | |||
| U37543.1 | - | 8 | 1503 | |||
| U37544.1 | - | 8 | 1503 | |||

Cd6
Genomic Map