siRNA Details
| Gene Symbol: Chil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009892.2 | NP_034022.2 | 607 | |||
| GenBank mRNA | AK137503.1 | - | 584 | |||
| AK154420.1 | - | 582 | ||||
| BC061154.1 | - | 582 | 30298306 | |||
| D87757.2 | - | 6 | 564 | |||
| M94584.2 | - | 576 | ||||
| U56900.1 | - | 2 | 151 | |||

Chil3
Genomic Map