siRNA Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089875.1 | - | 2 | 422 | ||
| AK089895.1 | - | 2 | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr3| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089875.1 | - | 2 | 422 | ||
| AK089895.1 | - | 2 | 293 | |||