siRNA Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009949.2 | NP_034079.2 | 1, 2 | 283 | ||
| GenBank mRNA | AK018752.1 | - | 1, 2 | 232 | ||
| AK150163.1 | - | 1, 2 | 255 | |||
| AK169038.1 | - | 1, 2 | 180 | |||
| BC138514.1 | - | 1, 2 | 218 | 8861534 | ||
| BC145859.1 | - | 1, 2 | 218 | 40131162 | ||
| BY325625.1 | - | 1, 2 | 283 | |||
| CD545813.1 | - | 1, 2 | 258 | |||
| U01163.1 | - | 1, 2 | 228 | |||

Cpt2
Genomic Map