siRNA Details
| Gene Symbol: Crygs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009967.2 | NP_034097.1 | 3 | 342 | ||
| GenBank mRNA | AF032995.1 | - | 3 | 338 | ||
| AK143060.1 | - | 3 | 339 | |||
| BC056452.1 | - | 3 | 318 | 6824006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crygs| Gene Symbol: Crygs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009967.2 | NP_034097.1 | 3 | 342 | ||
| GenBank mRNA | AF032995.1 | - | 3 | 338 | ||
| AK143060.1 | - | 3 | 339 | |||
| BC056452.1 | - | 3 | 318 | 6824006 | ||