siRNA Details
| Gene Symbol: Cmas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009908.2 | NP_034038.2 | 2 | 410 | ||
| GenBank mRNA | AJ006215.1 | - | 2 | 323 | ||
| AK155730.1 | - | 2 | 361 | |||
| BC031500.1 | - | 2 | 339 | 2647633 | ||
| BC063776.1 | - | 2 | 339 | 6398450 | ||

Cmas
Genomic Map