siRNA Details
Gene Symbol: Cox5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009942.2 | NP_034072.2 | 2, 3 | 279 | ||
GenBank mRNA | AK008339.1 | - | 2, 3 | 200 | ||
AK131673.1 | - | 2, 3 | 200 | |||
AK135174.1 | - | 2, 3 | 208 | |||
BC005564.1 | - | 219 | ||||
BC096048.1 | - | 2, 3 | 194 | 3465479 | ||
BC099474.1 | - | 2, 3 | 208 | 30933585 | ||
CJ075429.1 | - | 2, 3 | 284 | |||
X53157.1 | - | 2, 3 | 188 |
Gene Symbol: Gm11273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023228.1 | - | 207 |

More Information
Other Species Targeted

Panther Classification:
