siRNA Details
| Gene Symbol: Pcdha11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009960.1 | NP_034090.1 | 1 | 602 | ||
| GenBank mRNA | AB008182.1 | - | 1 | 495 | ||
| AK043705.1 | - | 1 | 710 | |||
| AK139436.1 | - | 1 | 748 | |||
| AK141960.1 | - | 1 | 749 | |||
| AY013758.1 | - | 1 | 602 | |||
| BC141349.1 | - | 1 | 628 | 9055961 | ||

Pcdha11
Genomic Map