siRNA Details
| Gene Symbol: Cryba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009965.2 | NP_034095.1 | 4 | 259 | ||
| GenBank mRNA | AJ239052.1 | - | 4 | 259 | ||
| AK084145.1 | - | 1 | 836 | |||
| BC107342.2 | - | 4 | 259 | 40055280 | ||
| J00378.1 | - | 1 | 32 | |||
| V00728.1 | - | 2 | 151 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_013056, X15143.

Cryba1
Genomic Map