siRNA Details
| Gene Symbol: Csf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009971.1 | NP_034101.1 | 5 | 868 | ||
| GenBank mRNA | BC120759.1 | - | 5 | 821 | 40129682 | |
| BC120761.1 | - | 5 | 821 | 40129684 | ||
| M13926.1 | - | 5 | 868 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf3| Gene Symbol: Csf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009971.1 | NP_034101.1 | 5 | 868 | ||
| GenBank mRNA | BC120759.1 | - | 5 | 821 | 40129682 | |
| BC120761.1 | - | 5 | 821 | 40129684 | ||
| M13926.1 | - | 5 | 868 | |||