siRNA Details
| Gene Symbol: Cst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009979.1 | NP_034109.1 | 1 | 234 | ||
| GenBank mRNA | AB017157.1 | - | 1 | 209 | ||
| AK006429.1 | - | 1 | 226 | |||
| BC048486.1 | - | 1 | 223 | 6772736 | ||
| Y18243.1 | - | 1 | 234 | |||

Cst9
Genomic Map