siRNA Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163159.1 | NP_001156631.1 | 3 | 475 | ||
NM_001163160.1 | NP_001156632.1 | 3 | 409 | |||
NM_009981.4 | NP_034111.1 | 3 | 311 | |||
GenBank mRNA | AK044585.1 | - | 3 | 312 | ||
AK076050.1 | - | 3 | 236 | |||
AK076830.1 | - | 3 | 410 | |||
BB642590.1 | - | 3 | 312 | |||
BC018313.1 | - | 2 | 194 | 4162053 | ||
L28956.1 | - | 3 | 280 | |||
Z12302.1 | - | 3 | 270 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
