siRNA Details
| Gene Symbol: Cyp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009994.1 | NP_034124.1 | 3 | 1708 | ||
| GenBank mRNA | AK016699.1 | - | 2 | 697 | ||
| AK017127.1 | - | 2 | 696 | |||
| AK041078.1 | - | 2 | 644 | |||
| AK137461.1 | - | 3 | 1627 | |||
| AK139368.1 | - | 3 | 1729 | |||
| U02479.1 | - | 1 | 272 | |||
| U03283.1 | - | 3 | 1708 | |||
| X78445.1 | - | 3 | 1531 | |||

Cyp1b1
Genomic Map