siRNA Details
| Gene Symbol: Cyp2c40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010004.2 | NP_034134.2 | 2 | 343 | ||
| GenBank mRNA | AF047727.1 | - | 2 | 257 | ||
| BC139471.1 | - | 2 | 343 | 8862493 | ||
| BC139472.1 | - | 2 | 343 | 8862494 | ||
| Gene Symbol: Cyp2c69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104525.1 | NP_001097995.1 | 2 | 313 | ||
| GenBank mRNA | BC145630.1 | - | 2 | 343 | ||

Cyp2c40
Genomic Map