siRNA Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009914.4 | NP_034044.3 | 2 | 2109 | ||
| GenBank mRNA | AK089875.1 | - | 2 | 2111 | ||
| AK089895.1 | - | 2 | 1981 | |||
| AK153518.1 | - | 2 | 2111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr3| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009914.4 | NP_034044.3 | 2 | 2109 | ||
| GenBank mRNA | AK089875.1 | - | 2 | 2111 | ||
| AK089895.1 | - | 2 | 1981 | |||
| AK153518.1 | - | 2 | 2111 | |||