siRNA Details
| Gene Symbol: Cnih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009919.2 | NP_034049.2 | 5 | 608 | ||
| GenBank mRNA | AF022811.1 | - | 5 | 599 | ||
| AK002293.1 | - | 5 | 576 | |||
| AK011200.1 | - | 5 | 561 | |||
| AK152432.1 | - | 5 | 559 | |||
| AK154479.1 | - | 5 | 559 | |||
| AK168154.1 | - | 5 | 559 | |||
| BC034868.1 | - | 5 | 568 | 2938052 | ||

Cnih1
Genomic Map