siRNA Details
| Gene Symbol: Cnr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134109.1 | - | 2 | 3508 | ||
| BC024052.1 | - | 2 | 3435 | 5098695 | ||
| X86405.1 | - | 2 | 3425 | |||
| X93168.1 | - | 2 | 3471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnr2| Gene Symbol: Cnr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134109.1 | - | 2 | 3508 | ||
| BC024052.1 | - | 2 | 3435 | 5098695 | ||
| X86405.1 | - | 2 | 3425 | |||
| X93168.1 | - | 2 | 3471 | |||