siRNA Details
| Gene Symbol: Cox7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009945.3 | NP_034075.2 | 2 | 285 | ||
| GenBank mRNA | AF037371.1 | - | 2 | 159 | ||
| AK007875.1 | - | 2 | 162 | |||
| AK011269.1 | - | 2 | 145 | |||
| AK019210.1 | - | 2 | 140 | |||
| AK153173.1 | - | 2 | 162 | |||
| BC010979.1 | - | 2 | 130 | 4217409 | ||
| CN670202.1 | - | 2 | 148 | |||
| CN838002.1 | - | 2 | 288 | |||
| X58486.1 | - | 2 | 117 | |||

Cox7a2
Genomic Map