siRNA Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256369.1 | NP_001243298.1 | 4 | 634 | ||
| NM_006641.3 | NP_006632.2 | 2 | 499 | |||
| NM_031200.2 | NP_112477.1 | 3 | 548 | |||
| XM_011533312.2 | XP_011531614.1 | 3 | 1261 | |||
| GenBank mRNA | AF145439.1 | - | 3 | 525 | ||
| AF145440.1 | - | 2 | 444 | |||
| AJ132337.1 | - | 3 | 427 | |||
| AK312824.1 | - | 4 | 589 | |||
| AK313366.1 | - | 3 | 502 | |||
| BC069678.1 | - | 3 | 509 | 7262627 | ||
| BC095516.1 | - | 1 | 356 | |||
| DB132476.1 | - | 2 | 494 | |||

CCR9
Genomic Map