siRNA Details
| Gene Symbol: Cyp2c38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010002.3 | NP_034132.2 | 9 | 2753 | ||
| GenBank mRNA | AK050051.1 | - | 9 | 2754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c38| Gene Symbol: Cyp2c38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010002.3 | NP_034132.2 | 9 | 2753 | ||
| GenBank mRNA | AK050051.1 | - | 9 | 2754 | ||