siRNA Details
| Gene Symbol: IER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003897.3 | NP_003888.2 | 2 | 294 | ||
| GenBank mRNA | AF039067.1 | - | 1 | 373 | ||
| AF083421.1 | - | 2 | 268 | |||
| AK222588.1 | - | 2 | 291 | |||
| AK314162.1 | - | 2 | 295 | |||
| BC000844.2 | - | 2 | 278 | 3457670 | ||
| BC005080.1 | - | 2 | 280 | 3617088 | ||
| BM915952.1 | - | 2 | 299 | |||
| BT006703.1 | - | 2 | 262 | |||
| DA893782.1 | - | 2 | 295 | |||
| S81914.1 | - | 2 | 280 | |||
| Y14551.1 | - | 2 | 273 | |||

IER3
Genomic Map