siRNA Details
| Gene Symbol: Dusp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010090.2 | NP_034220.2 | 2 | 590 | ||
| GenBank mRNA | AK088059.1 | - | 2 | 590 | ||
| AK134067.1 | - | 2 | 591 | |||
| BC048696.1 | - | 2 | 576 | 6511513 | ||
| L11330.1 | - | 2 | 545 | |||

Dusp2
Genomic Map