siRNA Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276387.1 | NP_001263316.1 | 4 | 1277 | ||
| NM_010095.6 | NP_034225.1 | 4 | 1272 | |||
| GenBank mRNA | AK053013.1 | - | 4 | 1273 | ||
| AK139262.1 | - | 4 | 984 | |||
| BC049188.1 | - | 3 | 571 | 6312260 | ||
| BC050922.1 | - | 3 | 794 | 6409737 | ||
| U71189.1 | - | 3 | 457 | |||
| U82441.1 | - | 3 | 326 | |||
| U92703.1 | - | 3 | 677 | |||

Ebf2
Genomic Map