siRNA Details
| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010139.3 | NP_034269.2 | 1, 2 | 218 | ||
| GenBank mRNA | AK137704.1 | - | 1, 2 | 200 | ||
| AK144202.1 | - | 1, 2 | 202 | |||
| BC140960.1 | - | 1, 2 | 174 | 9055572 | ||
| BY024029.1 | - | 1, 2 | 199 | |||
| CJ088701.1 | - | 1, 2 | 222 | |||
| U07634.1 | - | 1, 2 | 182 | |||
| X78339.1 | - | 1, 2 | 180 | |||

Epha2
Genomic Map