siRNA Details
| Gene Symbol: Ackr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010045.2 | NP_034175.2 | 2 | 1095 | ||
| GenBank mRNA | AF016584.1 | - | 2 | 1036 | ||
| BC005583.1 | - | 2 | 1094 | 3968984 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ackr1| Gene Symbol: Ackr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010045.2 | NP_034175.2 | 2 | 1095 | ||
| GenBank mRNA | AF016584.1 | - | 2 | 1036 | ||
| BC005583.1 | - | 2 | 1094 | 3968984 | ||