siRNA Details
| Gene Symbol: MSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193270.2 | NP_001180199.1 | 3, 4 | 382 | ||
| NM_001282174.1 | NP_001269103.1 | 3, 4 | 397 | |||
| NM_006800.3 | NP_006791.2 | 2, 3 | 367 | |||
| NM_078628.1 | NP_523352.1 | 3, 4 | 397 | |||
| NM_078629.3 | NP_523353.2 | 3, 4 | 397 | |||
| XM_005274440.2 | XP_005274497.1 | 3, 4 | 274 | |||
| XM_011545442.2 | XP_011543744.1 | 3, 4 | 351 | |||
| GenBank mRNA | AF117065.1 | - | 3, 4 | 397 | ||
| AK025642.1 | - | 2, 3 | 341 | |||
| AK289780.1 | - | 3, 4 | 319 | |||
| AK294255.1 | - | 3, 4 | 267 | |||
| AK300814.1 | - | 3, 4 | 355 | |||
| AK304419.1 | - | 2, 3 | 195 | |||
| BC031210.1 | - | 3, 4 | 330 | 5313214 | ||
| BI091178.1 | - | 3, 4 | 339 | |||
| CT002798.1 | - | 2, 3 | 431 | |||
| DA172776.1 | - | 3, 4 | 297 | |||
| DA788603.1 | - | 2, 3 | 369 | |||
| DA961602.1 | - | 1, 2 | 468 | |||
| DC395802.1 | - | 3, 4 | 355 | |||

MSL3
Genomic Map