siRNA Details
| Gene Symbol: Dyrk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037957.3 | NP_001033046.1 | 8 | 1293 | ||
| NM_001271370.1 | NP_001258299.1 | 8 | 1246 | |||
| NM_010092.2 | NP_034222.1 | 8 | 1293 | |||
| GenBank mRNA | AJ537610.1 | - | 8 | 1246 | ||
| AK148342.1 | - | 8 | 1273 | |||
| BC019545.1 | - | 8 | 1293 | 4511845 | ||
| Y18280.2 | - | 8 | 1282 | |||

Dyrk1b
Genomic Map