siRNA Details
| Gene Symbol: Eif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010120.5 | NP_034250.3 | 3 | 1500 | ||
| GenBank mRNA | AF026481.1 | - | 2 | 2321 | ||
| AK009026.1 | - | 3 | 1471 | |||
| AK013268.1 | - | 3 | 1499 | |||
| AK019451.1 | - | 3 | 1503 | |||
| AK150651.1 | - | 4 | 1426 | |||
| AK150673.1 | - | 3 | 1473 | |||
| AK150677.1 | - | 3 | 1473 | |||
| AK166368.1 | - | 1473 | ||||
| AK167333.1 | - | 3 | 1460 | |||
| BC027437.1 | - | 3 | 1463 | 4017054 | ||

Eif1a
Genomic Map