siRNA Details
| Gene Symbol: En1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010133.2 | NP_034263.2 | 2 | 2092 | ||
| GenBank mRNA | AK140408.1 | - | 2 | 1955 | ||
| L12703.1 | - | 2 | 1902 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

En1| Gene Symbol: En1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010133.2 | NP_034263.2 | 2 | 2092 | ||
| GenBank mRNA | AK140408.1 | - | 2 | 1955 | ||
| L12703.1 | - | 2 | 1902 | |||