siRNA Details
| Gene Symbol: Epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010149.3 | NP_034279.3 | 8 | 1245 | ||
| GenBank mRNA | AK010968.1 | - | 7 | 953 | ||
| AK139031.1 | - | 8 | 1144 | |||
| BC003953.1 | - | 8 | 1098 | |||
| BC046282.1 | - | 8 | 1175 | 6438591 | ||
| J04843.1 | - | 8 | 1127 | |||

Epor
Genomic Map