siRNA Details
| Gene Symbol: Eya4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010167.4 | NP_034297.2 | 8, 9 | 871 | ||
| GenBank mRNA | AK041625.1 | - | 8, 9 | 872 | ||
| BC120898.1 | - | 8, 9 | 855 | 40061597 | ||
| BC120899.1 | - | 8, 9 | 855 | 40061599 | ||
| Y17115.1 | - | 9, 10 | 982 | |||

Eya4
Genomic Map