siRNA Details
| Gene Symbol: GPR82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080817.4 | NP_543007.1 | 3 | 480 | ||
| XM_017029414.1 | XP_016884903.1 | 2 | 636 | |||
| GenBank mRNA | AJ319076.1 | - | 1 | 258 | ||
| AK122626.1 | - | 3 | 454 | |||
| AK313702.1 | - | 3 | 455 | |||
| AL832460.1 | - | 3 | 455 | |||
| BC070180.1 | - | 3 | 480 | 30351329 | ||
More Information
Set Membership: |
|

GPR82
Genomic Map