siRNA Details
| Gene Symbol: Fbln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010180.2 | NP_034310.2 | 3 | 329 | ||
| GenBank mRNA | AK035388.1 | - | 3 | 282 | ||
| AK083573.1 | - | 3 | 332 | |||
| AK085989.1 | - | 3 | 271 | |||
| AK086451.1 | - | 3 | 330 | |||
| AK137511.1 | - | 3 | 272 | |||
| AK159646.1 | - | 3 | 284 | |||
| AK183901.1 | - | 3 | 213 | |||
| BC007140.1 | - | 3 | 273 | 3495754 | ||
| EU543224.1 | - | 3 | 267 | |||
| X70854.1 | - | 3 | 326 | |||

Fbln1
Genomic Map