siRNA Details
| Gene Symbol: Fcer1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010185.4 | NP_034315.1 | 3 | 259 | ||
| GenBank mRNA | AK155600.1 | - | 3 | 216 | ||
| BC034163.1 | - | 199 | 5065647 | |||
| BY173505.1 | - | 3 | 259 | |||
| CK791068.1 | - | 3 | 209 | |||
| J05020.1 | - | 3 | 197 | |||

Fcer1g
Genomic Map