siRNA Details
| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010193.4 | NP_034323.1 | 1 | 531 | ||
| GenBank mRNA | AB022863.1 | - | 1 | 441 | ||
| AF064448.1 | - | 1 | 262 | |||
| AF178633.1 | - | 1 | 188 | |||
| AK032338.1 | - | 1 | 508 | |||
| AK132692.1 | - | 1 | 524 | |||
| AK149329.1 | - | 1 | 494 | |||
| AK154060.1 | - | 1 | 819 | |||
| AK208100.1 | - | 1 | 26 | |||
| BC068236.1 | - | 1 | 485 | 30544277 | ||

Fem1b
Genomic Map