siRNA Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010260.1 | NP_034390.1 | 5 | 742 | ||
| GenBank mRNA | AF077007.1 | - | 5 | 742 | ||
| AF109168.1 | - | 5 | 695 | |||
| AJ007970.1 | - | 5 | 735 | |||
| AK150138.1 | - | 5 | 678 | |||
| AK150146.1 | - | 1 | 44 | |||
| BC011336.1 | - | 5 | 731 | 3494311 | ||
| BC032882.1 | - | 5 | 757 | 1230883 | ||
| BC049141.1 | - | 5 | 719 | |||
| CT010249.1 | - | 4 | 572 | |||

Gbp2
Genomic Map