siRNA Details
| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010250.5 | NP_034380.1 | 9 | 1703 | ||
| GenBank mRNA | AK034095.1 | - | 9 | 1704 | ||
| AK039496.1 | - | 9 | 987 | |||
| AK141596.1 | - | 9 | 1563 | |||
| BC132331.1 | - | 9 | 1379 | 40130608 | ||
| BC137733.1 | - | 9 | 1382 | 8860752 | ||
| M63436.1 | - | 9 | 1376 | |||
| M86566.1 | - | 9 | 1071 | |||
| X61430.1 | - | 8 | 959 | |||

Gabra1
Genomic Map