siRNA Details
| Gene Symbol: Gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010271.2 | NP_034401.1 | 3 | 353 | ||
| GenBank mRNA | AK004565.1 | - | 3 | 358 | ||
| AK018733.1 | - | 3 | 359 | |||
| AK137134.1 | - | 3 | 360 | |||
| AK220445.1 | - | 3 | 347 | |||
| BC005756.1 | - | 3 | 320 | |||
| BC019391.1 | - | 3 | 353 | 4194849 | ||
| J02655.1 | - | 3 | 345 | |||

Gpd1
Genomic Map