siRNA Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010213.3 | NP_034343.2 | 6 | 1058 | ||
| GenBank mRNA | AF114382.1 | - | 6 | 1029 | ||
| AF134772.1 | - | 6 | 1028 | |||
| BC144903.1 | - | 6 | 1022 | 9053451 | ||
| BC145939.1 | - | 6 | 1022 | 40131242 | ||
| CB205184.1 | - | 4 | 715 | |||

Fhl3
Genomic Map