siRNA Details
| Gene Symbol: Gdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010267.3 | NP_034397.1 | 6 | 2310 | ||
| GenBank mRNA | AK083814.1 | - | 6 | 2311 | ||
| BC048177.1 | - | 6 | 2286 | 6825756 | ||
| BC051135.1 | - | 6 | 2275 | 6532839 | ||
| BC060212.1 | - | 6 | 2284 | |||
| Y17850.1 | - | 6 | 2298 | |||

Gdap1
Genomic Map