siRNA Details
| Gene Symbol: Gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010271.2 | NP_034401.1 | 8 | 2279 | ||
| GenBank mRNA | AK004565.1 | - | 8 | 2284 | ||
| AK018733.1 | - | 8 | 2285 | |||
| AK137134.1 | - | 8 | 2286 | |||
| AK220445.1 | - | 9 | 2238 | |||
| BC005756.1 | - | 8 | 2246 | |||
| BC019391.1 | - | 8 | 2279 | 4194849 | ||

Gpd1
Genomic Map