siRNA Details
| Gene Symbol: Gng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038637.1 | NP_001033726.1 | 5 | 318 | ||
| NM_001285908.1 | NP_001272837.1 | 4 | 278 | |||
| NM_001285909.1 | NP_001272838.1 | 6 | 385 | |||
| NM_001285910.1 | NP_001272839.1 | 3 | 425 | |||
| NM_001285911.1 | NP_001272840.1 | 4 | 293 | |||
| NM_010315.4 | NP_034445.1 | 4 | 466 | |||
| GenBank mRNA | AK003588.1 | - | 4 | 256 | ||
| AK012405.1 | - | 4 | 202 | |||
| AK036138.1 | - | 4 | 268 | |||
| AK075698.1 | - | 4 | 196 | |||
| AK158697.1 | - | 4 | 467 | |||
| AK160396.1 | - | 4 | 196 | |||
| BC021599.1 | - | 3 | 186 | 5150203 | ||
| BY108799.1 | - | 3 | 188 | |||
| BY245532.1 | - | 4 | 467 | |||
| CA324196.1 | - | 6 | 308 | |||
| U38496.1 | - | 2 | 53 | |||

Gng2
Genomic Map