siRNA Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010369.3 | NP_034499.3 | 2, 3 | 126 | ||
| GenBank mRNA | AK157851.1 | - | 2, 3 | 127 | ||
| AK162614.1 | - | 2, 3 | 111 | |||
| BC148192.1 | - | 2, 3 | 390 | 40045496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gypa| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010369.3 | NP_034499.3 | 2, 3 | 126 | ||
| GenBank mRNA | AK157851.1 | - | 2, 3 | 127 | ||
| AK162614.1 | - | 2, 3 | 111 | |||
| BC148192.1 | - | 2, 3 | 390 | 40045496 | ||